Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026496.4 | NP_080772.2 | 2 | 441 | ||
GenBank mRNA | AF411213.1 | - | 2 | 174 | ||
AK002633.1 | - | 2 | 442 | |||
BC055035.1 | - | 2 | 230 | 6306106 | ||
BY703140.1 | - | 2 | 442 |