Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHEK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005735.1 | NP_001005735.1 | 6 | 851 | ||
NM_001257387.1 | NP_001244316.1 | 6 | 836 | |||
NM_007194.3 | NP_009125.1 | 5 | 722 | |||
NM_145862.2 | NP_665861.1 | 5 | 722 | |||
XM_011529839.2 | XP_011528141.1 | 6 | 873 | |||
XM_011529840.2 | XP_011528142.1 | 6 | 875 | |||
XM_011529844.2 | XP_011528146.1 | 6 | 883 | |||
XM_011529845.2 | XP_011528147.1 | 4 | 429 | |||
XM_017028560.1 | XP_016884049.1 | 6 | 837 | |||
XM_017028561.1 | XP_016884050.1 | 2 | 81 | |||
GenBank mRNA | AB040105.1 | - | 4 | 650 | ||
AF086904.1 | - | 4 | 650 | |||
AF096279.1 | - | 4 | 650 | |||
AF174135.1 | - | 4 | 650 | |||
AF217975.1 | - | 6 | 1435 | |||
AJ131197.1 | - | 4 | 650 | |||
AK289360.1 | - | 5 | 701 | |||
AK290754.1 | - | 5 | 724 | |||
AY551297.1 | - | 5 | 779 | |||
AY551298.1 | - | 4 | 650 | |||
AY551299.1 | - | 4 | 650 | |||
AY551300.1 | - | 4 | 650 | |||
AY551301.1 | - | 4 | 540 | |||
AY551303.1 | - | 2 | 377 | |||
AY551305.1 | - | 5 | 764 | |||
BC004207.2 | - | 5 | 677 | 3532866 | ||
CR456418.1 | - | 6 | 850 |