Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marveld3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028584.3 | NP_082860.2 | 3 | 678 | ||
GenBank mRNA | AK007346.1 | - | 3 | 599 | ||
AW107118.1 | - | 4 | 498 | |||
BQ900249.1 | - | 3 | 319 |