Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fads1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146094.2 | NP_666206.1 | 12 | 1610 | ||
GenBank mRNA | AB072976.1 | - | 12 | 1525 | ||
AF061504.1 | - | 12 | 1511 | |||
AK033308.1 | - | 10 | 967 | |||
AK083959.1 | - | 12 | 1472 | |||
AK154367.1 | - | 12 | 1560 | |||
AK214855.1 | - | 1 | 167 | |||
AK218586.1 | - | 1 | 167 | |||
AK219592.1 | - | 1 | 167 | |||
BC013226.1 | - | 2 | 214 | |||
BC022139.1 | - | 12 | 1555 | 5100199 | ||
BC026831.1 | - | 12 | 1577 | 4239695 | ||
BC026848.1 | - | 12 | 1550 | 4196030 | ||
BC063053.1 | - | 12 | 1470 | 5696722 | ||
KF987079.1 | - | 12 | 1494 |