Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdpoz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148949.2 | NP_683751.2 | 2 | 611 | ||
GenBank mRNA | AF290198.1 | - | 2 | 596 | ||
BC125302.1 | - | 1 | 473 | 40129817 | ||
BC125304.1 | - | 1 | 473 | 40129819 |
Gene Symbol: Gm9125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163730.2 | NP_001157202.1 | 2 | 638 | ||
GenBank mRNA | AK033449.1 | - | 2 | 639 |