Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172689.3 | NP_766277.3 | 7 | 1058 | ||
GenBank mRNA | AK052595.1 | - | 3 | 312 | ||
AK078287.1 | - | 7 | 990 | |||
AK128929.1 | - | 7 | 993 | |||
AK155930.1 | - | 7 | 994 | |||
AY553221.1 | - | 7 | 934 | |||
BC027369.1 | - | 7 | 1037 | |||
BC106095.1 | - | 7 | 978 | |||
BC127163.1 | - | 7 | 924 | |||
BC152540.1 | - | 7 | 926 |