Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185092.1 | NP_001172021.1 | 5 | 592 | ||
NM_001185093.1 | NP_001172022.1 | 4 | 824 | |||
NM_001185094.1 | NP_001172023.1 | 5 | 593 | |||
NM_005600.2 | NP_005591.1 | 5 | 592 | |||
XM_005245214.2 | XP_005245271.1 | 5 | 943 | |||
XM_005245215.4 | XP_005245272.1 | 5 | 604 | |||
XM_005245216.3 | XP_005245273.1 | 5 | 615 | |||
XM_006711341.3 | XP_006711404.1 | 5 | 603 | |||
XM_017001371.1 | XP_016856860.1 | 5 | 619 | |||
GenBank mRNA | AF069987.1 | - | 5 | 563 | ||
AK296606.1 | - | 4 | 788 | |||
AK313323.1 | - | 5 | 559 | |||
BC046149.1 | - | 5 | 546 | 5764281 | ||
BI752623.1 | - | 5 | 593 | |||
BX416745.2 | - | 5 | 559 | |||
CB111935.1 | - | 5 | 592 | |||
CR541814.1 | - | 4 | 487 | |||
CR541846.1 | - | 4 | 487 |