Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYGM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF066859.1 | - | 3 | 371 | ||
AK299376.1 | - | 2 | 188 | |||
AK314474.1 | - | 3 | 375 | |||
BC029305.1 | - | 3 | 420 | |||
BC126392.1 | - | 3 | 770 | 8992108 | ||
BC130514.1 | - | 3 | 770 | 40146545 |