Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fstl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253719.1 | NP_001240648.1 | 4 | 359 | ||
NM_178673.4 | NP_848788.2 | 4 | 619 | |||
GenBank mRNA | AF374460.1 | - | 4 | 593 | ||
AK033652.1 | - | 4 | 610 | |||
AK046070.1 | - | 4 | 360 | |||
AK081152.1 | - | 4 | 610 | |||
AK138388.1 | - | 4 | 911 | |||
AK140611.1 | - | 4 | 622 | |||
BC075640.1 | - | 4 | 582 | 30545529 |