Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005624.3 | NP_005615.2 | 4, 5 | 447 | ||
XM_011528177.2 | XP_011526479.1 | 5, 6 | 648 | |||
XM_017027119.1 | XP_016882608.1 | 5, 6 | 614 | |||
XM_017027120.1 | XP_016882609.1 | 5, 6 | 543 | |||
XM_017027121.1 | XP_016882610.1 | 5, 6 | 489 | |||
GenBank mRNA | AB046579.1 | - | 2, 3 | 202 | ||
BC130561.1 | - | 3, 4 | 382 | 40146592 | ||
U86358.1 | - | 3, 4 | 336 |