Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005626.4 | NP_005617.2 | 3, 4 | 744 | ||
XM_011541951.2 | XP_011540253.1 | 3, 4 | 544 | |||
GenBank mRNA | AB209856.1 | - | 5, 6 | 6574 | ||
AK223444.1 | - | 3, 4 | 521 | |||
AK291509.1 | - | 3, 4 | 466 | |||
AK310528.1 | - | 7, 8 | 1232 | |||
BC002781.2 | - | 3, 4 | 468 | 3619538 | ||
BT007415.1 | - | 3, 4 | 371 | |||
L14076.1 | - | 3, 4 | 418 |