Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec24b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207209.2 | NP_997092.1 | 2 | 829 | ||
GenBank mRNA | BC046556.1 | - | 2 | 661 | ||
BC046776.1 | - | 2 | 829 | 5706007 | ||
BC057355.1 | - | 2 | 690 | 6833698 |