Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282189.1 | NP_001269118.1 | 5 | 337 | ||
NM_001282199.1 | NP_001269128.1 | 4 | 325 | |||
NM_005671.3 | NP_005662.1 | 5 | 448 | |||
XM_011544655.2 | XP_011542957.1 | 7 | 1525 | |||
XM_017013862.1 | XP_016869351.1 | 6 | 637 | |||
GenBank mRNA | AK296600.1 | - | 5 | 622 | ||
AY302140.1 | - | 5 | 312 | |||
BC020694.1 | - | 5 | 432 | 4772692 | ||
BI462115.1 | - | 5 | 453 | |||
BI755534.1 | - | 4 | 301 | |||
D83767.1 | - | 5 | 438 | |||
D83768.1 | - | 3 | 175 |