Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207636.2 | NP_997519.2 | 6 | 1064 | ||
GenBank mRNA | AK032733.1 | - | 6 | 1036 | ||
AK048586.1 | - | 6 | 1067 | |||
AK089380.1 | - | 6 | 1029 | |||
AK154790.1 | - | 6 | 833 | |||
BC144880.1 | - | 6 | 872 | |||
DQ192036.1 | - | 6 | 1065 |