Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252188.1 | NP_001239117.1 | 11 | 3564 | ||
NM_001252189.1 | NP_001239118.1 | 9 | 3320 | |||
NM_010148.3 | NP_034278.1 | 10 | 3356 | |||
GenBank mRNA | AK036331.1 | - | 9 | 3294 | ||
AK160782.1 | - | 9 | 3321 | |||
AK173093.1 | - | 11 | 3540 | |||
BC039138.1 | - | 10 | 3321 | 2647379 |