Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011986.4 | NP_036116.3 | 3 | 649 | ||
GenBank mRNA | AK154889.1 | - | 3 | 630 | ||
AK170502.1 | - | 4 | 364 | |||
BC017126.1 | - | 3 | 580 | 4485839 | ||
BQ745150.1 | - | 2 | 79 | |||
CX244321.1 | - | 3 | 592 | |||
CX568560.1 | - | 1 | 35 |