Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159986.1 | NP_001153458.1 | 7 | 909 | ||
NM_033354.3 | NP_203505.3 | 9 | 1156 | |||
GenBank mRNA | AK132144.1 | - | 7 | 933 | ||
AK173302.1 | - | 9 | 1137 | |||
BC059194.1 | - | 8 | 974 | 4222915 |