Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Txlnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138628.3 | NP_619534.2 | 9 | 1476 | ||
GenBank mRNA | AF422244.1 | - | 9 | 1413 | ||
AF422245.1 | - | 9 | 1413 | |||
AK084639.1 | - | 9 | 1477 | |||
AK085904.1 | - | 9 | 1372 | |||
AK137442.1 | - | 9 | 1391 | |||
BC141023.1 | - | 9 | 1389 | 9055635 |