Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thyn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144543.2 | NP_653126.1 | 5 | 494 | ||
GenBank mRNA | AB080370.1 | - | 4 | 418 | ||
AK021335.1 | - | 5 | 448 | |||
AK077639.1 | - | 5 | 464 | |||
BC016594.1 | - | 5 | 448 | 2631627 |