Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spsb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145134.3 | NP_660116.1 | 3 | 1938 | ||
GenBank mRNA | AF403039.2 | - | 3 | 1941 | ||
AK142195.1 | - | 3 | 1818 | |||
BC023083.1 | - | 2 | 1675 | 4953500 | ||
BC055705.1 | - | 3 | 1806 | 6412566 | ||
BC094332.1 | - | 3 | 2401 | 6817091 |