Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mypop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145579.3 | NP_663554.1 | 3 | 1983 | ||
GenBank mRNA | AF364868.1 | - | 4 | 1692 | ||
BC042748.1 | - | 2 | 1222 | |||
BC048544.1 | - | 1 | 133 | |||
M82905.1 | - | 1 | 243 | |||
M82906.1 | - | 1 | 105 |