Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110271.1 | NP_001103741.1 | 5 | 1000 | ||
NM_001110272.1 | NP_001103742.1 | 4 | 745 | |||
NM_145919.2 | NP_666031.2 | 6 | 876 | |||
GenBank mRNA | AB073619.1 | - | 6 | 824 | ||
AK003653.1 | - | 5 | 886 | |||
BC006879.1 | - | 6 | 833 | 2647263 | ||
CB055425.1 | - | 1 | 184 |