Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005588.2 | NP_005579.2 | 6 | 393 | ||
XM_011514628.1 | XP_011512930.1 | 5 | 450 | |||
XM_011514629.1 | XP_011512931.1 | 6 | 393 | |||
GenBank mRNA | AF478685.1 | - | 6 | 361 | ||
AK225212.1 | - | 6 | 383 | |||
AK290282.1 | - | 6 | 361 | |||
AK315246.1 | - | 6 | 365 | |||
AK315840.1 | - | 6 | 363 | |||
BC131714.1 | - | 6 | 364 | 40122874 | ||
BC143650.1 | - | 4 | 532 | |||
BC143651.1 | - | 5 | 443 | 9052165 | ||
DA432559.1 | - | 6 | 393 | |||
M82962.1 | - | 6 | 360 |