Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083903.1 | NP_001077372.1 | 2 | 506 | ||
NM_172205.3 | NP_757342.2 | 2 | 2060 | |||
GenBank mRNA | AA791703.1 | - | 2 | 304 | ||
AK003459.1 | - | 2 | 204 | |||
AY115494.1 | - | 2 | 2053 | |||
AY444556.1 | - | 2 | 445 | |||
BC029705.1 | - | 1 | 2242 | |||
BC051531.1 | - | 2 | 515 | |||
BC092350.1 | - | 2 | 435 | |||
BC139385.1 | - | 2 | 2052 | 8862407 |