Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP3CC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243974.1 | NP_001230903.1 | 15 | 2118 | ||
NM_001243975.1 | NP_001230904.1 | 13 | 2061 | |||
NM_005605.4 | NP_005596.2 | 14 | 2091 | |||
XM_017013610.1 | XP_016869099.1 | 14 | 1884 | |||
GenBank mRNA | AK299415.1 | - | 15 | 1952 | ||
AY007249.1 | - | 13 | 1667 | |||
BC004864.2 | - | 14 | 1978 | 3677098 | ||
S46622.1 | - | 13 | 1906 |