Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sned1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172463.4 | NP_766051.4 | 22 | 3136 | ||
GenBank mRNA | AJ584850.1 | - | 22 | 3192 | ||
AY169783.1 | - | 22 | 3191 | |||
BC145886.1 | - | 22 | 3136 | 40131189 |