Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039469.2 | NP_001034558.2 | 4 | 902 | ||
NM_001163296.1 | NP_001156768.1 | 4 | 902 | |||
NM_001163297.1 | NP_001156769.1 | 4 | 902 | |||
NM_004954.4 | NP_004945.4 | 4 | 902 | |||
NM_017490.3 | NP_059672.2 | 4 | 719 | |||
XM_011544789.1 | XP_011543091.1 | 5 | 793 | |||
XM_011544790.1 | XP_011543092.1 | 5 | 793 | |||
XM_011544791.1 | XP_011543093.1 | 5 | 793 | |||
XM_011544792.1 | XP_011543094.1 | 5 | 793 | |||
XM_011544793.1 | XP_011543095.1 | 5 | 793 | |||
XM_011544794.1 | XP_011543096.1 | 5 | 793 | |||
XM_011544795.1 | XP_011543097.1 | 5 | 793 | |||
XM_011544796.1 | XP_011543098.1 | 5 | 793 | |||
XM_011544797.1 | XP_011543099.1 | 5 | 793 | |||
XM_011544798.1 | XP_011543100.1 | 5 | 793 | |||
XM_017017310.1 | XP_016872799.1 | 4 | 875 | |||
XM_017017311.1 | XP_016872800.1 | 5 | 793 | |||
GenBank mRNA | AB188493.1 | - | 4 | 906 | ||
AB271694.1 | - | 3 | 224 | |||
AF387638.1 | - | 3 | 242 | |||
AK225766.1 | - | 4 | 737 | |||
AK290339.1 | - | 4 | 687 | |||
AK307858.1 | - | 5 | 538 | |||
BC008771.2 | - | 4 | 294 | |||
BC084540.1 | - | 4 | 361 | 4591688 | ||
BT007342.1 | - | 3 | 224 | |||
X97630.1 | - | 4 | 631 |