Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081238.2 | NP_001074707.1 | 4 | 826 | ||
NM_001289842.1 | NP_001276771.1 | 4 | 826 | |||
NM_001289843.1 | NP_001276772.1 | 3 | 732 | |||
GenBank mRNA | AK028653.1 | - | 4 | 800 | ||
AK154227.1 | - | 4 | 798 | |||
AK156701.1 | - | 4 | 745 | |||
AK157382.1 | - | 4 | 745 | |||
BC066139.1 | - | 3 | 674 |