Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165965.1 | NP_001159437.1 | 5, 6 | 1163 | ||
NM_177091.5 | NP_796065.2 | 6, 7 | 1421 | |||
GenBank mRNA | AK053989.1 | - | 6, 7 | 1284 | ||
AK054241.1 | - | 6, 7 | 1421 | |||
AK165075.1 | - | 3, 4 | 976 | |||
BC111898.1 | - | 5, 6 | 970 | 40061374 | ||
BC113188.1 | - | 6, 7 | 1227 |