Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C130071C03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029460.1 | - | 1723 | |||
AK048540.1 | - | 1724 | ||||
KF032589.1 | - | 2781 | ||||
KF032590.1 | - | 2706 |