Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177115.2 | NP_796089.1 | 3 | 542 | ||
GenBank mRNA | AK036622.1 | - | 3 | 510 | ||
AK041088.1 | - | 3 | 542 | |||
BC107232.1 | - | 3 | 326 | 40052073 | ||
BC107233.1 | - | 3 | 326 | 40052074 |