Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204466.1 | NP_001191395.1 | 16 | 2072 | ||
NM_001204467.1 | NP_001191396.1 | 17 | 2300 | |||
NM_001204468.1 | NP_001191397.1 | 17 | 2334 | |||
NM_005676.4 | NP_005667.2 | 17 | 2303 | |||
NM_152856.2 | NP_690595.1 | 16 | 2069 | |||
XM_005272677.4 | XP_005272734.1 | 17 | 2353 | |||
XM_005272678.4 | XP_005272735.1 | 16 | 1887 | |||
XM_005272679.4 | XP_005272736.1 | 16 | 1884 | |||
XM_006724563.1 | XP_006724626.1 | 13 | 1520 | |||
XM_011543989.2 | XP_011542291.1 | 9 | 1244 | |||
XM_017029884.1 | XP_016885373.1 | 16 | 2223 | |||
XM_017029885.1 | XP_016885374.1 | 16 | 1995 | |||
GenBank mRNA | AK025854.1 | - | 5 | 676 | ||
AK098115.1 | - | 2912 | ||||
AK292758.1 | - | 16 | 2023 | |||
AL137421.1 | - | 18 | 2192 | |||
BC000681.2 | - | 14 | 1525 | |||
BC003089.1 | - | 16 | 2008 | 3536058 | ||
BC004181.2 | - | 17 | 2256 | 2961094 | ||
BC008733.2 | - | 17 | 2288 | 3163064 | ||
BC024153.2 | - | 16 | 1918 | 3845258 | ||
BX537969.1 | - | 17 | 2285 | |||
D50912.1 | - | 17 | 2151 | |||
KU178411.1 | - | 15 | 1676 | |||
KU178412.1 | - | 16 | 1910 | |||
U35373.1 | - | 3 | 283 |