Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgea5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045615.1 | - | 1 | 797 | ||
AK054352.1 | - | 1 | 807 | |||
BC147259.1 | - | 1 | 396 | |||
BC147260.1 | - | 1 | 396 |