Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005692.4 | NP_005683.2 | 3 | 445 | ||
NM_007189.2 | NP_009120.1 | 3 | 445 | |||
XM_017011639.1 | XP_016867128.1 | 3 | 322 | |||
XM_017011640.1 | XP_016867129.1 | 3 | 407 | |||
GenBank mRNA | AF261091.1 | - | 3 | 302 | ||
AF261092.1 | - | 3 | 283 | |||
AL050291.1 | - | 2 | 237 | |||
BC001661.2 | - | 3 | 277 | 2988125 | ||
BT007451.1 | - | 2 | 191 | |||
CR987440.1 | - | 2 | 176 | |||
HY165500.1 | - | 3 | 449 |