Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL18BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005699.3 | NP_005690.2 | 4 | 2639 | ||
NM_173044.2 | NP_766632.2 | 6 | 1524 | |||
GenBank mRNA | AF110801.1 | - | 4 | 2639 | ||
AF215907.2 | - | 6 | 1513 |