Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177171.4 | NP_796145.2 | 27, 28 | 4416 | ||
GenBank mRNA | AK122493.1 | - | 16, 17 | 2420 | ||
AK161444.1 | - | 16, 17 | 2734 | |||
BC063265.1 | - | 3, 4 | 429 | |||
BC086656.1 | - | 26, 27 | 4241 | 30544145 |