Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmbrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177178.3 | NP_796152.2 | 6, 7 | 985 | ||
GenBank mRNA | AK036527.1 | - | 6, 7 | 1006 | ||
AK079450.1 | - | 6, 7 | 985 | |||
AK153728.1 | - | 6, 7 | 936 | |||
BC095971.1 | - | 6, 7 | 902 | 30108856 | ||
BC120727.1 | - | 6, 7 | 957 | 40129650 | ||
BC125421.1 | - | 6, 7 | 957 | 40129936 |