Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020778.4 | NP_065829.3 | 6 | 3065 | ||
XM_017022444.1 | XP_016877933.1 | 5 | 3063 | |||
XM_017022445.1 | XP_016877934.1 | 6 | 3114 | |||
GenBank mRNA | AB037751.1 | - | 1 | 14 | ||
AL832372.1 | - | 1 | 302 | |||
AL833276.1 | - | 1 | 381 | |||
AL833288.1 | - | 2 | 1747 | |||
AY044449.1 | - | 6 | 3065 |