Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lingo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177250.2 | NP_796224.1 | 2 | 1092 | ||
GenBank mRNA | AK040952.1 | - | 2 | 1092 | ||
AK042286.1 | - | 1793 | ||||
BC117866.1 | - | 2 | 1002 | 40095465 | ||
BC117867.1 | - | 2 | 1002 | 40095468 |