Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005731.3 | NP_005722.1 | 5 | 400 | ||
NM_152862.2 | NP_690601.1 | 6 | 495 | |||
XM_011510471.1 | XP_011508773.1 | 2 | 677 | |||
XM_017003113.1 | XP_016858602.1 | 5 | 369 | |||
XM_017003114.1 | XP_016858603.1 | 6 | 2778 | |||
XM_017003115.1 | XP_016858604.1 | 5 | 359 | |||
GenBank mRNA | AF006085.1 | - | 5 | 410 | ||
AF279893.1 | - | 4 | 252 | |||
AK092520.1 | - | 2 | 1457 | |||
AK093625.1 | - | 2 | 1448 | |||
AK290268.1 | - | 6 | 394 | |||
BC000590.2 | - | 6 | 358 | 3346685 | ||
BG282181.1 | - | 6 | 494 | |||
BG765898.1 | - | 6 | 438 | |||
BT006898.1 | - | 5 | 326 | |||
U50523.1 | - | 6 | 410 |