Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301173.1 | NP_001288102.1 | 12 | 2208 | ||
NM_177791.3 | NP_808459.1 | 13 | 2277 | |||
GenBank mRNA | AK076644.1 | - | 13 | 2269 | ||
BC092530.1 | - | 12 | 2208 | 6744085 | ||
BC125431.1 | - | 13 | 2269 | 40129946 | ||
BC125433.1 | - | 13 | 2269 | 40129948 |