Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038660.2 | NP_001033749.1 | 6 | 1922 | ||
NM_177802.3 | NP_808470.1 | 7 | 1479 | |||
GenBank mRNA | AK033481.1 | - | 7 | 1479 |