Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skint2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285963.1 | NP_001272892.1 | 4 | 461 | ||
NM_001285965.1 | NP_001272894.1 | 4 | 461 | |||
GenBank mRNA | AK040156.1 | - | 3 | 450 | ||
BC042786.1 | - | 4 | 628 | 3603407 | ||
EF494892.1 | - | 4 | 461 | |||
EF494893.1 | - | 3 | 461 | |||
EU099299.1 | - | 4 | 461 | |||
EU099300.1 | - | 4 | 461 | |||
EU650234.1 | - | 3 | 435 | |||
EU650235.1 | - | 3 | 435 |