Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABEPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174152.1 | NP_001167623.1 | 7 | 781 | ||
NM_001174153.1 | NP_001167624.1 | 6 | 628 | |||
NM_005833.3 | NP_005824.2 | 6 | 942 | |||
XM_005251640.4 | XP_005251697.1 | 6 | 854 | |||
XM_005251641.4 | XP_005251698.1 | 5 | 709 | |||
XM_005251642.4 | XP_005251699.1 | 5 | 700 | |||
XM_005251644.4 | XP_005251701.1 | 4 | 550 | |||
XM_011518120.2 | XP_011516422.1 | 6 | 857 | |||
XM_017014177.1 | XP_016869666.1 | 7 | 766 | |||
XM_017014178.1 | XP_016869667.1 | 6 | 668 | |||
XM_017014179.1 | XP_016869668.1 | 5 | 741 | |||
XM_017014180.1 | XP_016869669.1 | 5 | 515 | |||
XM_017014181.1 | XP_016869670.1 | 4 | 626 | |||
XM_017014182.1 | XP_016869671.1 | 5 | 451 | |||
XM_017014183.1 | XP_016869672.1 | 5 | 788 | |||
GenBank mRNA | AK290768.1 | - | 6 | 786 | ||
BC000503.2 | - | 6 | 919 | 2821471 | ||
BC047023.1 | - | 6 | 608 | 5396980 | ||
BC053541.1 | - | 6 | 751 | 5724458 | ||
BC065725.1 | - | 7 | 784 | 4102355 | ||
CR456837.1 | - | 5 | 612 | |||
CR541851.1 | - | 5 | 612 | |||
KU178507.1 | - | 5 | 612 | |||
KU178508.1 | - | 5 | 525 | |||
KU178509.1 | - | 4 | 463 | |||
KU178510.1 | - | 4 | 459 | |||
KU178511.1 | - | 3 | 297 | |||
Z97074.1 | - | 7 | 761 |