Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178778.3 | NP_848893.2 | 3 | 306 | ||
GenBank mRNA | AK044772.1 | - | 3 | 302 | ||
AK082408.1 | - | 4 | 408 | |||
AK083280.1 | - | 3 | 308 | |||
AK163712.1 | - | 3 | 310 | |||
BC132485.1 | - | 3 | 281 | 40130762 | ||
BC144986.1 | - | 4 | 382 | |||
BC144987.1 | - | 3 | 357 | |||
BC144989.1 | - | 4 | 382 |