Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skint4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178786.4 | NP_848901.2 | 3 | 481 | ||
GenBank mRNA | AK028550.1 | - | 3 | 532 | ||
AK028795.1 | - | 3 | 482 | |||
AK035748.1 | - | 3 | 399 | |||
AK162813.1 | - | 4 | 496 | |||
EF494897.1 | - | 3 | 280 | |||
EF494898.1 | - | 3 | 280 |