Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181820.2 | NP_861541.2 | 13 | 1989 | ||
GenBank mRNA | AK050105.1 | - | 12 | 1884 | ||
AY236493.1 | - | 13 | 1952 | |||
AY263162.1 | - | 13 | 1960 | |||
BC022758.1 | - | 5 | 679 | |||
BC040466.1 | - | 13 | 1901 | |||
CN719678.1 | - | 2 | 183 |