Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183175.4 | NP_898998.2 | 4 | 1695 | ||
GenBank mRNA | AK033673.1 | - | 1 | 1380 | ||
DQ002401.1 | - | 4 | 1695 |