Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206851.1 | NP_001193780.1 | 3 | 718 | ||
NM_183199.3 | NP_899022.2 | 3 | 718 | |||
GenBank mRNA | AK088109.1 | - | 3 | 719 | ||
BC111886.1 | - | 1 | 553 | 40060450 |