Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206851.1 | NP_001193780.1 | 3 | 1607 | ||
NM_183199.3 | NP_899022.2 | 4 | 1466 | |||
GenBank mRNA | AK088109.1 | - | 4 | 1467 | ||
BC111886.1 | - | 2 | 1301 | 40060450 |